Learn More
MSC Knaack Tool Tray, Steel, 16.625 in Length, Tan, Lift-Out
Catalog No. 5000536810
Encompass
The Model 41 lift-out tray is designed to suspend across the top opening of jobsite storage chests, making it easy to find small tools. It measures 8 inches wide by 16.625 inches long by 4 inches deep, preventing tools from piling up and allowing for easy lifting and removal. Its heavy-duty steel construction features a rugged tan powder-coat finish to prevent chips and scratches and increase durability. This tool tray is compatible with Models 32, 36, and 42.
  • Designed to suspend across the top of jobsite storage chest openings
  • Facilitates easy access and organization of small tools
  • Prevents tools from piling up
  • Lift-out design for convenient removal
  • Heavy-duty steel construction
  • Rugged tan powder-coat finish resists chips and scratches
  • Enhances durability
  • Portable
  • Compatible with Models 32, 36, and 42 portable tool chests
Catalog No. 50-005-36810 Supplier MSC Supplier No. 89761480
The Model 41 lift-out tray is designed to suspend across the top opening of jobsite storage chests, making it easy to find small tools. It measures 8 inches wide by 16.625 inches long by 4 inches deep, preventing tools from piling up and allowing for easy lifting and removal. Its heavy-duty steel construction features a rugged tan powder-coat finish to prevent chips and scratches and increase durability. This tool tray is compatible with Models 32, 36, and 42.
  • Designed to suspend across the top of jobsite storage chest openings
  • Facilitates easy access and organization of small tools
  • Prevents tools from piling up
  • Lift-out design for convenient removal
  • Heavy-duty steel construction
  • Rugged tan powder-coat finish resists chips and scratches
  • Enhances durability
  • Portable
  • Compatible with Models 32, 36, and 42 portable tool chests