Learn More
Cayman Chemical Kanamycin B, Purity: ≥95%, CAS: 4696-76-8, 100 mg
SDP
Catalog No. 502949784
Encompass_Preferred
An aminoglycoside antibiotic; active against S. aureus, E. coli, P. aeruginosa, and P. rettgeri(MICs = 0.78-25 µg/ml); binds to the ribosomal decoding A site to interfere with bacterial protein synthesis
Catalog No. 50-294-9784 Supplier Cayman Chemical Supplier No. 34561100 MG
An aminoglycoside antibiotic; active against S. aureus, E. coli, P. aeruginosa, and P. rettgeri(MICs = 0.78-25 µg/ml); binds to the ribosomal decoding A site to interfere with bacterial protein synthesis