Learn More
Cayman Chemical KnamycIn ELISA Stndard 1ea
SDP
Catalog No. 502942230
Encompass_Preferred
Kanamycin ELISA Standard has been tested and formulated to work exclusively with Cayman’s Kanamycin ELISA Kit (Item No. 502370)
Catalog No. 50-294-2230 Supplier Cayman Chemical Supplier No. 4005391 EA
Kanamycin ELISA Standard has been tested and formulated to work exclusively with Cayman’s Kanamycin ELISA Kit (Item No. 502370)