Learn More
Aviva Systems Biology Corp KNDC1 Antibody - C-terminal region (ARP60888_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60888_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KNDC1. It was validated on Western Blot. . Alternate names: C10orf23, FLJ16067, RASGEF2, bB439H18.3, RP13-439H18.3ApplicationWB. Species reactivity: Human
Catalog No. 50-287-197