Learn More
Sigma Aldrich Fine Chemicals Biosciences KNF LaboportR solid PTFE v1EA
Supplier: Sigma Aldrich Fine Chemicals Biosciences Z2882091EA
Quiet high performance diaphragm vacuum pumps can be used alone or as the center of a modular laboratory vacuum system. Replaces noisy rotary-vane pumps for vacuum distillation drying filtration rotary evaporation degassing of liquids and applications where water aspirators are used..nbspSolid PTFE heads.nbspPTFE coated diaphragm.nbspOil-free operation ensures pumped medium will stay pure.nbspNew multi-port valve system with Kalrez disks improves flow and reliability.nbspTwo stages.nbsp10 mm I.D. hose barbs on ports