Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KNF Neuberger LABOPORT™ Chemically Resistant Dry Vacuum Pumps: 60L/min. Flowrate
GSA_VA

Catalog No. 13880430 Shop All KNF Neuberger Products
Change view
Click to view available options
Flow Rate:
60 L/min.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Flow Rate
13-880-430 60 L/min.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13-880-430 Supplier KNF Neuberger Supplier No. 119655/049662
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Quiet, reliable operation without the hazards associated with handling messy pump oils or contaminated wastewater generated by aspirators

Handle corrosive and aggressive vapors without damage.

  • Oil-free
  • End vacuums to 1.5 torr

Feature solid PTFE heads and diaphragms to protect against corrosives and aggressive vapors. Multiport Kalrez™ valves withstand the ingestion of condensed vapors without the risk of damaging the pump or degrading performance.

Recommended for:

  • Rotary evaporators (13-878-27, -29, -35 and 13-880-430)
  • Vacuum concentrators (13-878-29, -35 and 13-880-430)
  • Gel dryers (13-878-27 and -29)
  • Vacuum ovens (13-878-27, -29, -35, and 13-880-430)
  • General vacuum applications (13-878-40, -42, -44, -46, 13-880-430 and -431)

Order Info

For 115 V/60 Hz; 230 V models are also available. For information contact your Fisher Scientific™ Customer Service Representative

1For 230 V/50 Hz models, contact your Fisher Scientific™ Customer Service Representative.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Max. Vacuum 1.5 torr
Electrical Requirements 115 V, 60 Hz
Dimensions (L x W x H) 331 x 291 x 278 mm (13.0 x 11.5 x 10.9 in.)
Model N860.3FTP
Flow Rate 60 L/min.
Frequency 60 Hz
Pump Type Diaphragm Vacuum Pump
Enclosure Rating IP20
Applications Rotary Evaporation
Maximum Pressure 1 bar
Voltage 115 V
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.