Learn More
Aviva Systems Biology Corp KNG1 Antibody - middle region (ARP45680_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45680_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KNG1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human KNG1. Alternate names: BDK, KNGApplicationWB. Species reactivity: Horse, Human, Mouse, Pig
Catalog No. 50-014-59098