Learn More
Fine Science Tools Dissecting Knives, Stainless Steel, Straight, 12.5 cm
SDP
Catalog No. NC9788024
Encompass
Product: Dissecting Knife - Straight/12.5cm/ Blade Shape: Straight/ Cutting Edge: 4mm/ Thickness: 0.5mm/ Alloy / Material: Stainless Steel
Catalog No. NC9788024 Supplier Fine Science Tools Supplier No. 1005512
Add to Cart
Add to Cart
Product: Dissecting Knife - Straight/12.5cm/ Blade Shape: Straight/ Cutting Edge: 4mm/ Thickness: 0.5mm/ Alloy / Material: Stainless Steel