Learn More
BYK Gardner KNIFE FILM CASTING 10.2 CM
SDP
Catalog No. 50894474
Encompass
Film Casting Knife, 10.2 cm film width, 0 - 3800 micron gap clearances
Catalog No. 50-894-474 Supplier BYK Gardner Supplier No. 2326
Film Casting Knife, 10.2 cm film width, 0 - 3800 micron gap clearances