Learn More
BYK Gardner KNIFE FILM CASTING 15.2 CM
SDP
Catalog No. 50894475
Encompass
The BYK-Gardner Film Casting Knife is an adjustable clearance filmapplicator. Its extended end plates confine the coating sampleduring drawdown. The micrometer adjusted gate allows clearance/ gap settings from 0 to 150 mils in 1 mil increments. Metricversions produce clearances / gaps of 0 to 3800 microns in 10micron increments.The applicator consists of two end plates joined by a bridge andan adjustable blade below the bridge. Two micrometers extendthrough the bridge and contact the upper edge of the blade,allowing it to be adjusted upward or downward to control thegap and ultimately the film thickness. The blade and end platesare constructed of 6.4 mm (1/4 in) aluminum. The end plates effectivelycontain the sample pool during the drawdown process. Blade Width 15.2cm Clearance/Gap 0-3800um. Dimensions (4x4x4.5in) Weight:(4lbs)
Catalog No. 50-894-475 Supplier BYK Gardner Supplier No. 2327
The BYK-Gardner Film Casting Knife is an adjustable clearance filmapplicator. Its extended end plates confine the coating sampleduring drawdown. The micrometer adjusted gate allows clearance/ gap settings from 0 to 150 mils in 1 mil increments. Metricversions produce clearances / gaps of 0 to 3800 microns in 10micron increments.The applicator consists of two end plates joined by a bridge andan adjustable blade below the bridge. Two micrometers extendthrough the bridge and contact the upper edge of the blade,allowing it to be adjusted upward or downward to control thegap and ultimately the film thickness. The blade and end platesare constructed of 6.4 mm (1/4 in) aluminum. The end plates effectivelycontain the sample pool during the drawdown process. Blade Width 15.2cm Clearance/Gap 0-3800um. Dimensions (4x4x4.5in) Weight:(4lbs)