Learn More
BYK Gardner KNIFE FILM CASTING 2
SDP
Catalog No. 50894494
Encompass
Film Casting Knife, 2 inch film width, 0 - 150 mil gap clearances
Catalog No. 50-894-494 Supplier BYK Gardner Supplier No. 4301
Film Casting Knife, 2 inch film width, 0 - 150 mil gap clearances