Learn More
Med Vet International Miltex Knife Handle, 8-1/4in, #4L, Holds Blades 20 Through 25, 4-11
Catalog No. 502643532
Encompass_Preferred
Miltex Knife Handle, 8-1/4in, #4L, Holds Blades 20 Through 25, 4-11
Catalog No. 50-264-3532 Supplier Med Vet International Supplier No. MIL411
Miltex Knife Handle, 8-1/4in, #4L, Holds Blades 20 Through 25, 4-11