Learn More
World Precision Instrument Stainless Steel Handle for Sapphire Blades
SDP
Catalog No. 50822441
Encompass
The Stainless Steel Scalpel Handle is for use with our Sapphire Blades. The 501317 version is for use with the regular sapphire blades (504074, 504075, and 504168), and the 501763 version is for use with angled sapphire blades. Since sapphires are one of the hardest materials available (second only to diamonds), they make super sharp blades for precise, clean cuts. The blades are non-magnetic, especially when combined with a titanium handle. Sapphire blades are non-conductive, which may be important if you are using electrical instruments. Sapphire blades are impervious to corrosion and extremely durable. The sapphire blades are sold separately. These handles are retractable, so you can protect the blade when it is not in use. The reusable scalpel handle is 13 cm (5.1") long.  They are constructed of surgical grade stainless steel and are autoclavable or suitable for chemical sterilization.
Catalog No. 50-822-441 Supplier World Precision Instrument Supplier No. 500317
The Stainless Steel Scalpel Handle is for use with our Sapphire Blades. The 501317 version is for use with the regular sapphire blades (504074, 504075, and 504168), and the 501763 version is for use with angled sapphire blades. Since sapphires are one of the hardest materials available (second only to diamonds), they make super sharp blades for precise, clean cuts. The blades are non-magnetic, especially when combined with a titanium handle. Sapphire blades are non-conductive, which may be important if you are using electrical instruments. Sapphire blades are impervious to corrosion and extremely durable. The sapphire blades are sold separately. These handles are retractable, so you can protect the blade when it is not in use. The reusable scalpel handle is 13 cm (5.1") long.  They are constructed of surgical grade stainless steel and are autoclavable or suitable for chemical sterilization.