Learn More
Wells Lamont Industrial™ Knifehandler White Glove
Catalog No. 18063989 Shop All Wells Lamont Industry Group Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
18-063-989 X-Large
18-063-986 Small
18-063-987 Medium
18-063-988 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-063-989 Supplier Wells Lamont Industrial™ Supplier No. 333376
Add to Cart
Add to Cart
Whizard™ Knifehandler ANSI A9 Level cut resistant glove. CFR 21 approved for food contact.
Whizard™ Knifehandler's single-end yarn construction offers the highest ratio of cut protection to thickness available.
  • Our unique patented combination of ultra-high strength fibers and stainless steel provides heavy duty protection from lacerations while maintaining softness, comfort and dexterity.
  • Knifehandler gloves have excellent thermal properties that keep hands warm and comfortable.
  • Breathable fabric increases circulation of air and helps to reduce perspiration.
Specifications
Construction Seamless Knit
Coating Material Uncoated
Color White
Cut Resistance Rating ANSI A9
Hand Ambidextrous
Latex Free Yes
Material Ultra-high Strength Fibers and Stainless Steel
Product Type Cut-resistant Gloves
Size X-Large
Gender Unisex
Glove Cuff Style Elasticated
Lining Unlined
Puncture Rating ASTM F2878 LEVEL 0
Thumb Style Straight