Learn More
Grainger KNIPEX Bolt Cutter, Steel
Catalog No. NC2884622
Encompass_Preferred
The KNIPEX 61KP84 bolt cutter is a 24-inch steel bolt cutter designed for cutting bolts and rods up to 3/4 inches in diameter. It features chrome vanadium electric steel jaws with a cutting edge hardness of 62 HRC. The bolt cutter has ergonomic, 2-component handles with soft grips for comfort and reduced hand fatigue. It has a maximum cutting force of over 4 tons. The overall length is 24 inches and it weighs 9.5 lbs. This heavy-duty bolt cutter from KNIPEX is made in Germany.
Catalog No. NC2884622 Supplier Grainger Supplier No. 61KP84
The KNIPEX 61KP84 bolt cutter is a 24-inch steel bolt cutter designed for cutting bolts and rods up to 3/4 inches in diameter. It features chrome vanadium electric steel jaws with a cutting edge hardness of 62 HRC. The bolt cutter has ergonomic, 2-component handles with soft grips for comfort and reduced hand fatigue. It has a maximum cutting force of over 4 tons. The overall length is 24 inches and it weighs 9.5 lbs. This heavy-duty bolt cutter from KNIPEX is made in Germany.