Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Valutek™ Knit Polyester Strands Floor Mop
Description
- This product is recommended for use in a cleanroom Class 100-1,000 (ISO 5-6) critical environment.
- The mop head is compatible with a wide range of chemicals. This makes it ideal for single use of wet mopping of floors in controlled environments, such as lab and process area floor cleaning.
- Laundered and packaged in a cleanroom, all Valutek™ mop heads are tested and manufactured in ISO compliant facilities under Valutek™ inspection and strict process control to ensure Valutek™ quality standards and product specifications.
Specifications
Specifications
| Dimensions | 7 x 16 in. |
| Sterility | Non-sterile |
| Description | 5 in. Strands |
| Material | Polyester |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?