Learn More
Sigma Aldrich Fine Chemicals Biosciences Knitted reactor coil L x I.D. 5 m x 0.5 mm, volume 1.0 mL |
Catalog No. NC2294652
Encompass
Knitted reactor coil L x I.D. 5 m x 0.5 mm, volume 1.0 mL |
Catalog No. NC2294652 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 57405
Knitted reactor coil L x I.D. 5 m x 0.5 mm, volume 1.0 mL |