Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ KnockOut™ DMEM
GREENER_CHOICE

Catalog No. 10829018
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-829-018 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 10-829-018 Supplier Gibco™ Supplier No. 10829018
Season of Savings
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

KnockOut D-MEM is a basal medium optimized for growth of undifferentiated embryonic and induced pluripotent stem cells. The osmolarity is optimized to approximate that of mouse embryonic tissue.

KnockOut D-MEM is a basal medium optimized for growth of undifferentiated embryonic and induced pluripotent stem cells. The osmolarity is optimized to approximate that of mouse embryonic tissue. It is designed specifically to improve the morphology of ES cells. The osmolality of KnockOut D-MEM has been reduced to better mimic the natural environment of embryonic tissue. When used together, KnockOutD-MEM and KnockOut SR significantly reduce ES cell differentiation as compared to ES Cell Qualified FBS and traditional D-MEM.

KnockOut D-MEM contains no L-glutamine.

KnockOut™ D-MEM is a basal medium optimized for growth of undifferentiated embryonic and induced pluripotent stem cells (1). The osmolarity is optimized to approximate that of mouse embryonic tissue.

Contains no L-glutamine.

TRUSTED_SUSTAINABILITY

Specifications

Cell Type Stem Cells (Embryonic), Stem Cells (iPS - Induced Pluripotent Stem)
Form Liquid
Product Type KnockOut DMEM
With Additives High Glucose, Phenol Red
Without Additives No Glutamine
Manufacturing Quality cGMP for medical devices, 21 CFR Part 820 and ISO 13485
Product Line KnockOut
Quantity 500 mL
Shipping Condition Room Temperature
Content And Storage Store in refrigerator (2–8°C) and protect from light.
Can KnockOut SR be used to culture mouse ESCs in feeder-free conditions?

Yes, mouse embryonic stem cells can be cultured using KnockOut SR under feeder-free conditions. Typically, cells are plated at a higher seeding density, in the presence of LIF, on a 0.1% gelatin layer in place of feeder cells.

Can KnockOut SR be used to culture mouse ESCs on feeder cells?

Yes, KnockOut SR was designed to culture mouse ES cells in undifferentiated conditions.

How do I characterize human embryonic stem ( ES) cells?

Human ES cells are generally characterized by their typical morphology (they grow as tightly packed clusters of small cells with high ratio of nucleus to cytoplasm); surface marker expression; RT-PCR detection of stem cell-specific gene expression (such as Oct3/4, Sox2, and Nanog); alkaline phosphatase staining, and telomerase activity assay. The most commonly used ES specific surface markers include stage-specific embryonic antigens SSEA-3 and SSEA-4 for human ES cells. Other ES-specific surface antigens also include TRA-1-60 and TRA-1-81. (Science 282:1145 (1998).

How are human embryonic stem (ES) cells derived?

Human ES cells are derived from human blastocyst inner cell masses, isolated by immunosurgery with rabbit antiserum to BeWO cells (a human trophoblast cell line) (Science 282:1145 (1998)).

What are ES cells?

Embryonic stem (ES) cells are derived from the early mammalian embryo and are capable of unlimited, undifferentiated proliferation in vitro while maintaining their potential to differentiate into a wide of range of adult tissues including germ cells. The pluripotency of the ES cells is normally demonstrated in vitro by inducing ES cells to differentiate into embryoid bodies and checking lineage-specific markers for differentiated cells in three body layers (endo, meso, and ectoderm), or injecting them into immunodeficient mice and determining the cell types produced in the teratomas.

WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.