Learn More
Cliawaived Inc 16 Panel Know Drug Test Cups
SDP
Catalog No. NC1698844
Encompass_Preferred
(25/Pack) 16 Panel Urine Test Cups (AMP 500, BAR 200, BUP 10, BZO 200, COC 150, ETG 500, FEN 25, K2, mAMP 500, MDMA 500, MTD 300, OPI 300, OXY 100, PCP 25, TRAM 200, THC 50 + CRE/OX/pH). This cups tests for many illicit substances and prescription drugs. New 16 panel cup is the most comprehensive on the market and also includes tests for Tramadol, Alcohol (ETG metabolite offers 80 hours of detection for Alcohol), Fentanyl and K2 Spice. For workplace and insurance testing (not FDA cleared or CLIA waived).
This item is not returnable. View return policy
Catalog No. NC1698844 Supplier Cliawaived Inc Supplier No. WP16KDTCA
(25/Pack) 16 Panel Urine Test Cups (AMP 500, BAR 200, BUP 10, BZO 200, COC 150, ETG 500, FEN 25, K2, mAMP 500, MDMA 500, MTD 300, OPI 300, OXY 100, PCP 25, TRAM 200, THC 50 + CRE/OX/pH). This cups tests for many illicit substances and prescription drugs. New 16 panel cup is the most comprehensive on the market and also includes tests for Tramadol, Alcohol (ETG metabolite offers 80 hours of detection for Alcohol), Fentanyl and K2 Spice. For workplace and insurance testing (not FDA cleared or CLIA waived).