Learn More
Med Vet International Miltex Vantage Knowles Bandage Scissors, 5.5in.
Catalog No. 502829579
Encompass_Preferred
The Miltex Vantage V95-560 Knowles bandage scissors are made from high quality Japanese stainless steel and measure 5.5" in length. They have a finger ring handle and straight blades. One of the tips is blunt and the other is sharp.
Catalog No. 50-282-9579 Supplier Med Vet International Supplier No. MILV95560
The Miltex Vantage V95-560 Knowles bandage scissors are made from high quality Japanese stainless steel and measure 5.5" in length. They have a finger ring handle and straight blades. One of the tips is blunt and the other is sharp.