Learn More
Medchemexpress LLC KNT-127 | 1256921-89-7 | 99.6% | 1 MG
SDP
Catalog No. 5000231897
Encompass_Preferred
KNT-127 is a selective, brain-penetrant δ-opioid receptor agonist used in neuroscience research. It exhibits high affinity for the δ receptor and functional bias toward cAMP signaling, and has demonstrated antidepressant- and anxiolytic-like effects in animal models.
  • Selective δ-opioid receptor agonist with high affinity (Ki δ ≈ 0.16 nM).
  • Biased signaling toward cAMP over beta-arrestin pathways.
  • Brain-penetrant compound suitable for in vivo studies.
  • High purity solid (99.6%) available in milligram quantities.
  • Recommended storage: powder at -20°C for long-term stability.
Catalog No. 50-002-31897 Supplier Medchemexpress LLC Supplier No. HY1205111MG
KNT-127 is a selective, brain-penetrant δ-opioid receptor agonist used in neuroscience research. It exhibits high affinity for the δ receptor and functional bias toward cAMP signaling, and has demonstrated antidepressant- and anxiolytic-like effects in animal models.
  • Selective δ-opioid receptor agonist with high affinity (Ki δ ≈ 0.16 nM).
  • Biased signaling toward cAMP over beta-arrestin pathways.
  • Brain-penetrant compound suitable for in vivo studies.
  • High purity solid (99.6%) available in milligram quantities.
  • Recommended storage: powder at -20°C for long-term stability.