Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Knt-127 | 1256921-89-7 | 99.6% | 1 ML
SDP

Catalog No. 5000234528
Encompass_Preferred

KNT-127 is a selective, brain-penetrant δ-opioid receptor (DOR) agonist supplied as a 10 mM solution in DMSO. It acts as a biased ligand favoring cAMP signaling over β-arrestin pathways and is used in preclinical research on opioid receptor pharmacology and neurological models. Not for human or veterinary use.

  • Selective δ-opioid receptor agonist with high potency
  • Biased signaling that favors cAMP over β-arrestin pathways
  • Supplied as a 10 mM solution in DMSO, 1 mL
  • High purity suitable for research (99.6%)
  • Molecular formula C24H24N2O2; molecular weight 372.46 g/mol

Catalog No. 50-002-34528 Supplier Medchemexpress LLC Supplier No. HY12051110MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

KNT-127 is a selective, brain-penetrant δ-opioid receptor (DOR) agonist supplied as a 10 mM solution in DMSO. It acts as a biased ligand favoring cAMP signaling over β-arrestin pathways and is used in preclinical research on opioid receptor pharmacology and neurological models. Not for human or veterinary use.

  • Selective δ-opioid receptor agonist with high potency
  • Biased signaling that favors cAMP over β-arrestin pathways
  • Supplied as a 10 mM solution in DMSO, 1 mL
  • High purity suitable for research (99.6%)
  • Molecular formula C24H24N2O2; molecular weight 372.46 g/mol

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.