Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Knt-127 | 1256921-89-7 | 99.6% | 10 MG
SDP

Catalog No. 5000232715
Encompass_Preferred

KNT-127 is a selective δ-opioid receptor agonist used in neuroscience research, reported to produce antidepressant-like and antinociceptive effects in preclinical models. CAS: 1256921-89-7. It is supplied as an off-white to light yellow solid with high purity and is suitable for in vitro and in vivo studies.

  • Selective δ-opioid receptor agonist.
  • Reported antidepressant-like and antinociceptive effects in animal models.
  • High purity (99.6%).
  • Molecular weight 372.46 g/mol.
  • Soluble in DMSO at 25 mg/mL.
  • Powder stable when stored at -20°C for extended periods.
  • Suitable for in vitro pharmacology and in vivo behavioral studies.
  • Available in small research quantities for laboratory use.

Catalog No. 50-002-32715 Supplier Medchemexpress LLC Supplier No. HY12051110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

KNT-127 is a selective δ-opioid receptor agonist used in neuroscience research, reported to produce antidepressant-like and antinociceptive effects in preclinical models. CAS: 1256921-89-7. It is supplied as an off-white to light yellow solid with high purity and is suitable for in vitro and in vivo studies.

  • Selective δ-opioid receptor agonist.
  • Reported antidepressant-like and antinociceptive effects in animal models.
  • High purity (99.6%).
  • Molecular weight 372.46 g/mol.
  • Soluble in DMSO at 25 mg/mL.
  • Powder stable when stored at -20°C for extended periods.
  • Suitable for in vitro pharmacology and in vivo behavioral studies.
  • Available in small research quantities for laboratory use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.