Learn More
Medchemexpress LLC Knt-127 | 1256921-89-7 | 99.6% | 25 MG
SDP
Catalog No. 5000232902
Encompass_Preferred
KNT-127 is a selective, blood-brain barrier penetrant δ-opioid receptor (DOR) agonist used in neuroscience research; it preferentially activates cAMP signaling with reduced β-arrestin recruitment.
  • Selective δ-opioid receptor agonist with reported Ki = 0.16 nM
  • Blood-brain barrier penetrant, suitable for in vivo studies
  • Biased ligand favoring cAMP signaling over β-arrestin pathways
  • High reported purity (99.6%) appropriate for research applications
  • Available in small pack sizes, including 25 mg
Catalog No. 50-002-32902 Supplier Medchemexpress LLC Supplier No. HY12051125MG
KNT-127 is a selective, blood-brain barrier penetrant δ-opioid receptor (DOR) agonist used in neuroscience research; it preferentially activates cAMP signaling with reduced β-arrestin recruitment.
  • Selective δ-opioid receptor agonist with reported Ki = 0.16 nM
  • Blood-brain barrier penetrant, suitable for in vivo studies
  • Biased ligand favoring cAMP signaling over β-arrestin pathways
  • High reported purity (99.6%) appropriate for research applications
  • Available in small pack sizes, including 25 mg