Learn More
Medchemexpress LLC KNT-127 | 1256921-89-7 | 99.6% | 5 MG
SDP
Catalog No. 5000234403
Encompass_Preferred
KNT-127 is a selective, blood-brain barrier penetrant δ-opioid receptor agonist used in neurological research. It exhibits high affinity for the δ receptor and biased signaling that favors cAMP pathways, with reported effects in preclinical models.
  • Selective δ-opioid receptor agonist.
  • Blood-brain barrier penetrant for central nervous system studies.
  • Biased ligand favoring cAMP signaling over β-arrestin.
  • High purity suitable for in vivo and in vitro research applications.
  • Available in small-mass packages for dosing flexibility.
Catalog No. 50-002-34403 Supplier Medchemexpress LLC Supplier No. HY1205115MG
KNT-127 is a selective, blood-brain barrier penetrant δ-opioid receptor agonist used in neurological research. It exhibits high affinity for the δ receptor and biased signaling that favors cAMP pathways, with reported effects in preclinical models.
  • Selective δ-opioid receptor agonist.
  • Blood-brain barrier penetrant for central nervous system studies.
  • Biased ligand favoring cAMP signaling over β-arrestin.
  • High purity suitable for in vivo and in vitro research applications.
  • Available in small-mass packages for dosing flexibility.