Learn More
Medchemexpress LLC KNT-127 | 1256921-89-7 | 99.6% | 50 MG
SDP
Catalog No. 5000234402
Encompass_Preferred
KNT-127 is a selective, blood-brain barrier-penetrant δ-opioid receptor (DOR) agonist used in preclinical neuroscience research. It displays high affinity for DOR (Ki = 0.16 nM), biased signaling toward cAMP over β-arrestin pathways, and has been reported to increase dopamine and L-glutamate release in key brain regions. For research use only.
  • Selective δ-opioid receptor agonist with high DOR potency.
  • Biased ligand favoring cAMP signaling over β-arrestin recruitment.
  • Blood-brain barrier penetrant, suitable for central nervous system studies.
  • High chemical purity (99.6%) and off-white to light yellow solid form.
  • Available in small solid quantities and DMSO solution formats for in vitro and in vivo work.
Catalog No. 50-002-34402 Supplier Medchemexpress LLC Supplier No. HY12051150MG
Add to Cart
Add to Cart
Request a Quote
KNT-127 is a selective, blood-brain barrier-penetrant δ-opioid receptor (DOR) agonist used in preclinical neuroscience research. It displays high affinity for DOR (Ki = 0.16 nM), biased signaling toward cAMP over β-arrestin pathways, and has been reported to increase dopamine and L-glutamate release in key brain regions. For research use only.
  • Selective δ-opioid receptor agonist with high DOR potency.
  • Biased ligand favoring cAMP signaling over β-arrestin recruitment.
  • Blood-brain barrier penetrant, suitable for central nervous system studies.
  • High chemical purity (99.6%) and off-white to light yellow solid form.
  • Available in small solid quantities and DMSO solution formats for in vitro and in vivo work.