Learn More
McMaster-Carr 91746A119, Hex Socket Head Cap Screw, M6 x 1.0 mm Thread, 25 mm Long, Alloy Steel, Black Oxide Finish
Supplier:  McMaster-Carr 91746A119
Encompass
The McMaster-Carr 91746A119 is a stainless steel hex head cap screw with a thread size of 1/4"-20, a length of 1-1/2", and a tensile strength of 170,000 psi. It has a flat point for use in thin materials and its 18-8 stainless steel construction provides strength and offers good corrosion resistance. This cap screw meets ASME B18.3 specifications and is made in the USA.
Catalog No. NC2339013