Learn More
TARGETMOL CHEMICALS INC Ko 143 10MG
SDP
Catalog No. 502880287
Encompass_Preferred
Also available in 1 mL, 2 mg, 5 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Ko 143 is a selective inhibitor of ATP-binding cassette sub-family G member 2 (ABCG2; BCRP). Purity 99.89%
Catalog No. 50-288-0287 Supplier TARGETMOL CHEMICALS INC Supplier No. TQ018610MG
Also available in 1 mL, 2 mg, 5 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Ko 143 is a selective inhibitor of ATP-binding cassette sub-family G member 2 (ABCG2; BCRP). Purity 99.89%