Learn More
Medchemexpress LLC HY-10010 10mg Medchemexpress, Ko 143 CAS:461054-93-3 Purity:>98%
SDP
Catalog No. 502028607
Encompass_Preferred
Medchemexpress, HY-10010 10mg Ko 143 CAS:461054-93-3 Ko 143 is a potent and selective ATP-binding cassette subfamily G member 2 (ABCG2/BCRP) inhibitor. Ko 143 displays >200-fold selectivity over P-gp and MRP-1 transporters[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8607 Supplier Medchemexpress LLC Supplier No. HY1001010MG
Medchemexpress, HY-10010 10mg Ko 143 CAS:461054-93-3 Ko 143 is a potent and selective ATP-binding cassette subfamily G member 2 (ABCG2/BCRP) inhibitor. Ko 143 displays >200-fold selectivity over P-gp and MRP-1 transporters[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.