Learn More
Apexbio Technology LLC Ko 143 461054-93-3 50mg
SDP
Catalog No. 5000093742
Encompass_Preferred
Ko 143 (CAS 461054-93-3) is a selective inhibitor of the breast cancer resistance protein (BCRP/ABCG2) an ATP-binding cassette transporter implicated in multidrug resistance in cancer cells By targeting BCRP Ko 143 effectively blocks BCRP-mediated drug efflux as demonstrated by an EC90 of 26 nM in cell-based assays This compound exhibits over 200-fold selectivity for BCRP compared to P-glycoprotein and MRP1 In vitro Ko 143 reverses BCRP-mediated resistance to chemotherapeutics such as topotecan and mitoxantrone enhancing their potency up to tenfold in relevant cancer cell lines Ko 143 has been used in murine models to increase systemic exposure of BCRP substrates supporting its utility in studying drug resistance mechanisms and improving chemotherapeutic efficacy in preclinical research
Catalog No. 50-000-93742 Supplier Apexbio Technology LLC Supplier No. A353350
Ko 143 (CAS 461054-93-3) is a selective inhibitor of the breast cancer resistance protein (BCRP/ABCG2) an ATP-binding cassette transporter implicated in multidrug resistance in cancer cells By targeting BCRP Ko 143 effectively blocks BCRP-mediated drug efflux as demonstrated by an EC90 of 26 nM in cell-based assays This compound exhibits over 200-fold selectivity for BCRP compared to P-glycoprotein and MRP1 In vitro Ko 143 reverses BCRP-mediated resistance to chemotherapeutics such as topotecan and mitoxantrone enhancing their potency up to tenfold in relevant cancer cell lines Ko 143 has been used in murine models to increase systemic exposure of BCRP substrates supporting its utility in studying drug resistance mechanisms and improving chemotherapeutic efficacy in preclinical research