Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ko 143 | 461054-93-3 | MFCD19053160 | 100.0% | 469.6 g/mol | C26H35N3O5 | 1 ML
SDP

Catalog No. 5000215456
Encompass_Preferred

Potent, selective ABCG2 (BCRP) inhibitor used to probe transporter function and multidrug resistance. Shows over 200-fold selectivity versus P-gp and MRP-1; supplied as a 10 mM solution in 1 mL for laboratory use.

  • Potent and selective ABCG2 (BCRP) inhibitor for transporter studies.
  • Displays greater than 200-fold selectivity over P-gp and MRP-1.
  • Supplied as a 10 mM solution in 1 mL for convenient dosing.
  • Molecular weight 469.6 g/mol and formula C26H35N3O5.
  • CAS number 461054-93-3 for unambiguous identification.

Catalog No. 50-002-15456 Supplier Medchemexpress LLC Supplier No. HY1001010MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Potent, selective ABCG2 (BCRP) inhibitor used to probe transporter function and multidrug resistance. Shows over 200-fold selectivity versus P-gp and MRP-1; supplied as a 10 mM solution in 1 mL for laboratory use.

  • Potent and selective ABCG2 (BCRP) inhibitor for transporter studies.
  • Displays greater than 200-fold selectivity over P-gp and MRP-1.
  • Supplied as a 10 mM solution in 1 mL for convenient dosing.
  • Molecular weight 469.6 g/mol and formula C26H35N3O5.
  • CAS number 461054-93-3 for unambiguous identification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.