Learn More
Medchemexpress LLC Ko 143 | 461054-93-3 | MFCD19053160 | 99.9% | 469.57 g·mol⁻¹ | C26H35N3O5 | 50 MG
SDP
Catalog No. 5000215660
Encompass_Preferred
Ko 143 is a potent, selective inhibitor of the ATP-binding cassette subfamily G member 2 (ABCG2/BCRP) used as a research reagent to investigate transporter-mediated drug efflux and multidrug resistance. It exhibits nanomolar activity in biochemical and cell-based assays and shows substantial selectivity versus other transporters. Supplied as a research-grade small molecule for laboratory use only.
  • Potent ABCG2 inhibition at nanomolar concentrations.
  • High selectivity over P-gp and MRP-1.
  • Suitable for studies of transporter activity and drug resistance.
  • Supplied in milligram quantities for laboratory experiments.
  • Stable as powder when stored appropriately at low temperature.
Catalog No. 50-002-15660 Supplier Medchemexpress LLC Supplier No. HY1001050MG
Ko 143 is a potent, selective inhibitor of the ATP-binding cassette subfamily G member 2 (ABCG2/BCRP) used as a research reagent to investigate transporter-mediated drug efflux and multidrug resistance. It exhibits nanomolar activity in biochemical and cell-based assays and shows substantial selectivity versus other transporters. Supplied as a research-grade small molecule for laboratory use only.
  • Potent ABCG2 inhibition at nanomolar concentrations.
  • High selectivity over P-gp and MRP-1.
  • Suitable for studies of transporter activity and drug resistance.
  • Supplied in milligram quantities for laboratory experiments.
  • Stable as powder when stored appropriately at low temperature.