Learn More
Abcam Ko-143, BCRP inhibitor, 5MG
Catalog No. 502704851
Encompass_Preferred
MW 469.6 Da. Potent, selective BCRP inhibitor (EC₅₀ values are 23 and 26 nM respectively for the export of mitoxantrone (ab145668, ab141041) and topotecan (ab120388) in cancer cell lines). Non-toxic fumitremorgin C (ab144258) analog. Shows chemopotentiating effects in vivo. .
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4851 Supplier Abcam Supplier No. AB1461985MG
MW 469.6 Da. Potent, selective BCRP inhibitor (EC₅₀ values are 23 and 26 nM respectively for the export of mitoxantrone (ab145668, ab141041) and topotecan (ab120388) in cancer cell lines). Non-toxic fumitremorgin C (ab144258) analog. Shows chemopotentiating effects in vivo. .
The product is subject to the following: Abcam Restricted Use Statement