Learn More
Medchemexpress LLC HY-112181 10mg Medchemexpress, KO-947 CAS:1695533-89-1 Purity:>98%
SDP
Catalog No. 501969054
Encompass_Preferred
Medchemexpress, HY-112181 10mg KO-947 CAS:1695533-89-1 KO-947 is a potent and selective inhibitor of ERK1/2 kinases with potential clinical utility in MAPK pathway dysregulated tumors. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9054 Supplier Medchemexpress LLC Supplier No. HY11218110MG
Medchemexpress, HY-112181 10mg KO-947 CAS:1695533-89-1 KO-947 is a potent and selective inhibitor of ERK1/2 kinases with potential clinical utility in MAPK pathway dysregulated tumors. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.