Learn More
ABclonal Technology [KO Validated] ANXA1 Rabbit pAb
SDP
Catalog No. 5000115166
Encompass_Preferred
This gene encodes a membrane-localized protein that binds phospholipids. This protein inhibits phospholipase A2 and has anti-inflammatory activity. Loss of function or expression of this gene has been detected in multiple tumors.
Catalog No. 50-001-15166 Supplier ABclonal Technology Supplier No. A11181000UL
This gene encodes a membrane-localized protein that binds phospholipids. This protein inhibits phospholipase A2 and has anti-inflammatory activity. Loss of function or expression of this gene has been detected in multiple tumors.