Learn More
ABclonal Technology KO Validated CDK4 Rabbit
SDP
Catalog No. 5000005026
Encompass_Preferred
The protein encoded by this gene is a member of the Ser Thr protein kinase family This protein is highly similar to the gene products of S cerevisiae cdc28 and S pombe cdc2 It is a catalytic subunit of the protein kinase complex that is important for cell cycle G1 phase progression The activity of this kinase is restricted to the G1-S phase which is controlled by the regulatory subunits D-type cyclins and CDK inhibitor p16(INK4a) This kinase was shown to be responsible for the phosphorylation of retinoblastoma gene product (Rb) Mutations in this gene as well as in its related proteins including D-type cyclins p16(INK4a) and Rb were all found to be associated with tumorigenesis of a variety of cancers Multiple polyadenylation sites of this gene have been reported
Catalog No. 50-000-05026 Supplier ABclonal Technology Supplier No. A23521PM20UL
The protein encoded by this gene is a member of the Ser Thr protein kinase family This protein is highly similar to the gene products of S cerevisiae cdc28 and S pombe cdc2 It is a catalytic subunit of the protein kinase complex that is important for cell cycle G1 phase progression The activity of this kinase is restricted to the G1-S phase which is controlled by the regulatory subunits D-type cyclins and CDK inhibitor p16(INK4a) This kinase was shown to be responsible for the phosphorylation of retinoblastoma gene product (Rb) Mutations in this gene as well as in its related proteins including D-type cyclins p16(INK4a) and Rb were all found to be associated with tumorigenesis of a variety of cancers Multiple polyadenylation sites of this gene have been reported