Learn More
ABclonal Technology [KO Validated] CHRAC1 Rabbit pAb
SDP
Supplier:  ABclonal Technology A148961000UL
Encompass_Preferred
CHRAC1 is a histone-fold protein that interacts with other histone-fold proteins to bind DNA in a sequence-independent manner. These histone-fold protein dimers combine within larger enzymatic complexes for DNA transcription, replication, and packaging.
Catalog No. 50-001-06332