Learn More
ABclonal Technology [KO Validated] GIT1 Rabbit pAb
SDP
Catalog No. 5000106086
Encompass_Preferred
Enables gamma-tubulin binding activity. Involved in positive regulation of microtubule nucleation and regulation of cytokinesis. Located in several cellular components, including focal adhesion, microtubule cytoskeleton, and mitochondrion. Implicated in attention deficit hyperactivity disorder. Biomarker of Huntingtons disease.
Catalog No. 50-001-06086 Supplier ABclonal Technology Supplier No. A15437500UL
Enables gamma-tubulin binding activity. Involved in positive regulation of microtubule nucleation and regulation of cytokinesis. Located in several cellular components, including focal adhesion, microtubule cytoskeleton, and mitochondrion. Implicated in attention deficit hyperactivity disorder. Biomarker of Huntingtons disease.