Learn More
ABclonal Technology KO Validated Hexokinase II
SDP
Catalog No. 5000006839
Encompass_Preferred
Hexokinases phosphorylate glucose to produce glucose-6-phosphate the first step in most glucose metabolism pathways This gene encodes hexokinase 2 the predominant form found in skeletal muscle It localizes to the outer membrane of mitochondria Expression of this gene is insulin-responsive and studies in rat suggest that it is involved in the increased rate of glycolysis seen in rapidly growing cancer cells
Catalog No. 50-000-06839 Supplier ABclonal Technology Supplier No. A22319100UL
Hexokinases phosphorylate glucose to produce glucose-6-phosphate the first step in most glucose metabolism pathways This gene encodes hexokinase 2 the predominant form found in skeletal muscle It localizes to the outer membrane of mitochondria Expression of this gene is insulin-responsive and studies in rat suggest that it is involved in the increased rate of glycolysis seen in rapidly growing cancer cells