Learn More
ABclonal Technology KO Validated KRAS Rabbit mAb
SDP
Catalog No. 5000005735
Encompass_Preferred
This gene a Kirsten ras oncogene homolog from the mammalian ras gene family encodes a protein that is a member of the small GTPase superfamily A single amino acid substitution is responsible for an activating mutation The transforming protein that results is implicated in various malignancies including lung adenocarcinoma mucinous adenoma ductal carcinoma of the pancreas and colorectal carcinoma Alternative splicing leads to variants encoding two isoforms that differ in the C-terminal region
Catalog No. 50-000-05735 Supplier ABclonal Technology Supplier No. A2338220UL
This gene a Kirsten ras oncogene homolog from the mammalian ras gene family encodes a protein that is a member of the small GTPase superfamily A single amino acid substitution is responsible for an activating mutation The transforming protein that results is implicated in various malignancies including lung adenocarcinoma mucinous adenoma ductal carcinoma of the pancreas and colorectal carcinoma Alternative splicing leads to variants encoding two isoforms that differ in the C-terminal region