Learn More
ABclonal Technology KO Validated PHD2/EGLN1
SDP
Supplier:  ABclonal Technology A2125220UL
Encompass_Preferred
The protein encoded by this gene catalyzes the post-translational formation of 4-hydroxyproline in hypoxia-inducible factor (HIF) alpha proteins HIF is a transcriptional complex that plays a central role in mammalian oxygen homeostasis This protein functions as a cellular oxygen sensor and under normal oxygen concentration modification by prolyl hydroxylation is a key regulatory event that targets HIF subunits for proteasomal destruction via the von Hippel-Lindau ubiquitylation complex Mutations in this gene are associated with erythrocytosis familial type 3 (ECYT3)
Catalog No. 50-000-06598