Learn More
ABclonal Technology KO Validated PRMT4/CARM1
SDP
Catalog No. 5000007702
Encompass_Preferred
This gene belongs to the protein arginine methyltransferase (PRMT) family The encoded enzyme catalyzes the methylation of guanidino nitrogens of arginyl residues of proteins The enzyme acts specifically on histones and other chromatin-associated proteins and is involved in regulation of gene expression The enzyme may act in association with other proteins or within multi-protein complexes and may play a role in cell type-specific functions and cell lineage specification A related pseudogene is located on chromosome 9
Catalog No. 50-000-07702 Supplier ABclonal Technology Supplier No. A21106100UL
This gene belongs to the protein arginine methyltransferase (PRMT) family The encoded enzyme catalyzes the methylation of guanidino nitrogens of arginyl residues of proteins The enzyme acts specifically on histones and other chromatin-associated proteins and is involved in regulation of gene expression The enzyme may act in association with other proteins or within multi-protein complexes and may play a role in cell type-specific functions and cell lineage specification A related pseudogene is located on chromosome 9