Learn More
ABclonal Technology KO Validated SMARCB1/SNF5
SDP
Catalog No. 5000006764
Encompass_Preferred
The protein encoded by this gene is part of a complex that relieves repressive chromatin structures allowing the transcriptional machinery to access its targets more effectively The encoded nuclear protein may also bind to and enhance the DNA joining activity of HIV-1 integrase This gene has been found to be a tumor suppressor and mutations in it have been associated with malignant rhabdoid tumors Alternatively spliced transcript variants have been found for this gene
Catalog No. 50-000-06764 Supplier ABclonal Technology Supplier No. A2346620UL
The protein encoded by this gene is part of a complex that relieves repressive chromatin structures allowing the transcriptional machinery to access its targets more effectively The encoded nuclear protein may also bind to and enhance the DNA joining activity of HIV-1 integrase This gene has been found to be a tumor suppressor and mutations in it have been associated with malignant rhabdoid tumors Alternatively spliced transcript variants have been found for this gene