Learn More
MSC Kobalt Air Compressor Accessory Kit, 18-piece, 1/4 inch NPT, Brass Fittings
Catalog No. 5000507148
Encompass
This 18-piece air compressor accessory kit provides essential tools to maximize air compressor utility. It includes a blow gun, tire gauge, dual-chuck inflator, and various brass quick-connect fittings. Ideal for inflating tires, cleaning, and connecting air tools, it features durable, corrosion-resistant brass fittings and a zippered storage bag for organization.
  • Corrosion-resistant 1/4-inch NPT brass plugs and couplers for easy connections with most air compressors, air tools, and hoses.
  • Zippered storage bag for organized and accessible storage.
  • Comprehensive kit includes a blow gun, tire gauge, dual-chuck inflator, inflation needles, tapered nozzle, adapters, and additional attachments for diverse uses.
  • Ideal for inflating tires, sports equipment, air mattresses, and cleaning hard-to-reach spots.
  • Features a safety nozzle to reduce the risk of injury.
Catalog No. 50-005-07148 Supplier MSC Supplier No. 77681377
This 18-piece air compressor accessory kit provides essential tools to maximize air compressor utility. It includes a blow gun, tire gauge, dual-chuck inflator, and various brass quick-connect fittings. Ideal for inflating tires, cleaning, and connecting air tools, it features durable, corrosion-resistant brass fittings and a zippered storage bag for organization.
  • Corrosion-resistant 1/4-inch NPT brass plugs and couplers for easy connections with most air compressors, air tools, and hoses.
  • Zippered storage bag for organized and accessible storage.
  • Comprehensive kit includes a blow gun, tire gauge, dual-chuck inflator, inflation needles, tapered nozzle, adapters, and additional attachments for diverse uses.
  • Ideal for inflating tires, sports equipment, air mattresses, and cleaning hard-to-reach spots.
  • Features a safety nozzle to reduce the risk of injury.