Learn More
TARGETMOL CHEMICALS INC KOBE0065 50MG
SDP
Catalog No. 502581068
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 100 mg 200 mg 500 mg and bulk. Please contact Fisher for quotes. Kobe0065 is a novel and effective small-molecule compound inhibiting Ras–Raf interaction by SBDD; shows potent activity to competitively inhibit the binding of H-Ras·GTP to c-Raf-1 RBD with a Ki value of 46 ± 13 uM. purity: 99%
Catalog No. 50-258-1068 Supplier TARGETMOL CHEMICALS INC Supplier No. T187650MG
Also available in 5 mg 10 mg 25 mg 100 mg 200 mg 500 mg and bulk. Please contact Fisher for quotes. Kobe0065 is a novel and effective small-molecule compound inhibiting Ras–Raf interaction by SBDD; shows potent activity to competitively inhibit the binding of H-Ras·GTP to c-Raf-1 RBD with a Ki value of 46 ± 13 uM. purity: 99%