Learn More
Medchemexpress LLC HY-15717 50mg Medchemexpress, Kobe2602 CAS:454453-49-7 Purity:>98%
SDP
Catalog No. 502081777
Encompass_Preferred
Medchemexpress, HY-15717 50mg Kobe2602 CAS:454453-49-7 Kobe2602 is a Ras-Raf interaction inhibitor. Kobe2602 inhibits the binding of H-Ras·GTP to c-Raf-1 RBD with a Ki of 149 μM. Kobe2602 has antitumor activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1777 Supplier Medchemexpress LLC Supplier No. HY1571750MG
Medchemexpress, HY-15717 50mg Kobe2602 CAS:454453-49-7 Kobe2602 is a Ras-Raf interaction inhibitor. Kobe2602 inhibits the binding of H-Ras·GTP to c-Raf-1 RBD with a Ki of 149 μM. Kobe2602 has antitumor activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.