Learn More
ATCC Kocuria rhizophila; Strain PCI 1001
Catalog No. 502382833
Encompass
Kocuria rhizophila strain FDA strain PCI 1001 was isolated from soil. This whole-genome sequenced bacterial strain is recommended as a test microorganism in USP chapters <71> and <81> and it can be used as a quality control strain in sterility testing and susceptibility disc testing.
Catalog No. 50-238-2833 Supplier ATCC Supplier No. 9341
Kocuria rhizophila strain FDA strain PCI 1001 was isolated from soil. This whole-genome sequenced bacterial strain is recommended as a test microorganism in USP chapters <71> and <81> and it can be used as a quality control strain in sterility testing and susceptibility disc testing.