Learn More
Sigma Aldrich Fine Chemicals Biosciences KOD OneTM PCR Master Mix R
Catalog No. 502904789
Encompass
KOD One(TM) PCR master Mix is a ready-to-use 2 x PCR master mix containing a novel genetically modified KOD DNA polymerase (UKOD) along with a new elongation accelerator enabling fast PCR with an extension time of 5 sec/kb for template DNA 10kb. This master mix has greater efficiency and flexibility than conventional PCR enzymes with an ability to amplify from crude specimens such as blood and mouse cell lysates. In addition to crude samples it can be used on templates containing uracils (dU). The master mix can use primers containing both inosines (dI) and dU for amplification.This hot start KOD polymerase is antibody-inactivated with two types of antibodies that prevent the 5-3 polymerase activity and 3-5 exonuclease activity. Upon heat-activation the 3-5 proof-reading ability of the enzyme generates blunt-end PCR products.
Catalog No. 50-290-4789 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. KMM101NV5X1ML
KOD One(TM) PCR master Mix is a ready-to-use 2 x PCR master mix containing a novel genetically modified KOD DNA polymerase (UKOD) along with a new elongation accelerator enabling fast PCR with an extension time of 5 sec/kb for template DNA 10kb. This master mix has greater efficiency and flexibility than conventional PCR enzymes with an ability to amplify from crude specimens such as blood and mouse cell lysates. In addition to crude samples it can be used on templates containing uracils (dU). The master mix can use primers containing both inosines (dI) and dU for amplification.This hot start KOD polymerase is antibody-inactivated with two types of antibodies that prevent the 5-3 polymerase activity and 3-5 exonuclease activity. Upon heat-activation the 3-5 proof-reading ability of the enzyme generates blunt-end PCR products.