Learn More
TOYOBO USA INC KOD sybr qPCR Mix, High efficient real-time PCR master Mix
Catalog No. NC1788632
Encompass
Quantitative analysis can be achieved using long targets, up to 2kb. Efficient for GC-rich targets. Effective amplification can be achieved using crude samples.
Catalog No. NC1788632 Supplier TOYOBO USA INC Supplier No. QKD201
Quantitative analysis can be achieved using long targets, up to 2kb. Efficient for GC-rich targets. Effective amplification can be achieved using crude samples.