Learn More
Electron Microscopy Sciences Kodak Rapid Fixer, 1 Gal Kit
SDP
Supplier:  Electron Microscopy Sciences 74312
Encompass_Preferred
A rapid-acting hardening fixer for automated and tray processing. Complete fixer. Kit makes 1 gal: 32oz solution A and 3.6 oz solution B.
Catalog No. 50-268-10